2-Fluor-α-methyl-4-biphenylessigsäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Ansaid, 2-(2-Fluoro-[1,1'-biphenyl]-4-yl)propanoic acid, Flurbiprofen, Froben, Antadys

M-Clarity™ Qualitätsstufe = MQ100

F8514-1G F8514-25G F8514-5G
SIALF8514-1GEA 124 EUR
SIALF8514-1G SIALF8514-25G SIALF8514-5G
2-Fluor-α-methyl-4-biphenylessigsäure, Sigma-Aldrich®
2-Fluor-α-methyl-4-biphenylessigsäure
Formel: C₁₅H₁₃FO₂
Molecular Weight: 244.265 g/mol
MDL: MFCD00079303
CAS-Nummer: 5104-49-4
EINECS: 225-827-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR