5-Fluor-5'-deoxyuridin (Doxifluridin), Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ200

F8791-100MG F8791-250MG F8791-25MG
SIALF8791-100MGEA 229 EUR
SIALF8791-100MG SIALF8791-250MG SIALF8791-25MG
5-Fluor-5'-deoxyuridin (Doxifluridin), Sigma-Aldrich®
5-Fluor-5'-deoxyuridin (Doxifluridin)
Formel: C₉H₁₁FN₂O₅
MDL: MFCD00866530
CAS-Nummer: 3094-09-5
EINECS: 221-440-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR