Flash point standards, Pensky Martens closed cup, ASTM D 93, Supelco®
Lieferant: Merck
FPPMCC1-3X80ML FPPMCC4-3X80ML FPPMCC3-3X80ML FPPMCC5-3X80ML FPPMCC6-3X80ML FPPMCC2-3X80ML
SIALFPPMCC1-3X80MLEA 254 EUR
SIALFPPMCC1-3X80ML SIALFPPMCC4-3X80ML SIALFPPMCC3-3X80ML SIALFPPMCC5-3X80ML SIALFPPMCC6-3X80ML SIALFPPMCC2-3X80ML
Flash point standards, Pensky Martens closed cup, ASTM D 93, Supelco®
Standards Mineralölindustrie-Standards
Procedure A is applicable to distillate fuels - diesels, bio-diesel blends, kerosene, heating oil, turbine fuels and lubricating oils. These UKAS certified standards are specifically designed to be used for regular instrument performance checks.
Standards: These products are certified reference materials (CRMs) certified to International Standards BS EN ISO / IEC 17025 and ISO 17034 under UKAS accreditation.