Flash point standards, Pensky Martens closed cup, ASTM D 93, Supelco®
Lieferant: Merck
FPPMCC1-3X80ML
FPPMCC4-3X80ML
FPPMCC3-3X80ML
FPPMCC5-3X80ML
FPPMCC6-3X80ML
FPPMCC2-3X80ML
SIALFPPMCC1-3X80MLEA
254
EUR
SIALFPPMCC1-3X80ML
SIALFPPMCC4-3X80ML
SIALFPPMCC3-3X80ML
SIALFPPMCC5-3X80ML
SIALFPPMCC6-3X80ML
SIALFPPMCC2-3X80ML
Flash point standards, Pensky Martens closed cup, ASTM D 93, Supelco®
Standards
Mineralölindustrie-Standards
Procedure A is applicable to distillate fuels - diesels, bio-diesel blends, kerosene, heating oil, turbine fuels and lubricating oils. These UKAS certified standards are specifically designed to be used for regular instrument performance checks.
Standards: These products are certified reference materials (CRMs) certified to International Standards BS EN ISO / IEC 17025 and ISO 17034 under UKAS accreditation.