Guillard’s (F/2) marine water enrichment solution, Sigma-Aldrich®
Lieferant: Merck
G0154-500ML
SIALG0154-500MLEA 108 EUR
SIALG0154-500ML
Guillard’s (F/2) marine water enrichment solution, Sigma-Aldrich®
Medien für die Mikrobiologie Additive und Supplemente für die Mikrobiologie
Guillard’s (F/2) marine water enrichment solution 50X, is a concentrate of major nutrients, trace metals and vitamins that can be diluted in filtered seawater to support reconstitution of Guillard’s (F/2) marine enrichment medium.
Guillard’s (F/2) marine enrichment medium is a widely used enriched seawater medium for growing costal marine algae (diatoms).
Reconstitution: Use at 20 ml/L of seawater.