3',4'-Dimethoxy-2'-hydroxyacetophenon, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Gallacetophenone 3',4'-dimethyl ether

G1411-1G
SIALG1411-1GEA 52.4 EUR
SIALG1411-1G
3',4'-Dimethoxy-2'-hydroxyacetophenon, Sigma-Aldrich®
3',4'-Dimethoxy-2'-hydroxyacetophenon
Formel: C₁₀H₁₂O₄
MDL: MFCD00016684
CAS-Nummer: 5396-18-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR