1,4-Dioxan-2,5-dion, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Glycolide, 1,4-Dioxane-2,5-dione

M-Clarity™ Qualitätsstufe = MQ200

G1796-25G
SIALG1796-25GEA 175 EUR
SIALG1796-25G
1,4-Dioxan-2,5-dion, Sigma-Aldrich®
1,4-Dioxan-2,5-dion
Formel: C₄H₄O₄
Molecular Weight: 116,07 g/mol
MDL: MFCD00081108
CAS-Nummer: 502-97-6
EINECS: 207-954-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR