Gellan Gummi, Sigma-Aldrich®

Lieferant: Merck


MQ200

G1910-1KG G1910-250G G1910-5KG
SIALG1910-1KGEA 753 EUR
SIALG1910-1KG SIALG1910-250G SIALG1910-5KG
Gellan Gummi, Sigma-Aldrich®
Gellan Gummi
Lagertemperatur: Raumtemperatur
MDL: MFCD00131909
CAS-Nummer: 71010-52-1
EINECS: 275-117-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR