Glycinbenzylester Hydrochlorid (H-Gly-OBzl.HCl), Sigma-Aldrich®

Lieferant: Merck

Synonyme: H-Gly-OBzl·HCl, H-DL-Gly-OBzl hydrochloride, Benzyl glycinate hydrochloride, H-DL-Gly-OBzl.HCl

M-Clarity™ Qualitätsstufe = MQ200

G3267-5G
SIALG3267-5GEA 130 EUR
SIALG3267-5G
Glycinbenzylester Hydrochlorid (H-Gly-OBzl.HCl), Sigma-Aldrich®
Glycinbenzylester Hydrochlorid (H-Gly-OBzl.HCl)
Formel: C₉H₁₂ClNO₂
Molecular Weight: 201.65 g/mol
MDL: MFCD00035442
CAS-Nummer: 2462-31-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR