di-Natrium-β-glycerophosphat Hydrat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: ortho-Phosphorsäure(2-hydroxy-1-(hydroxymethyl)ethyl)ester Dinatriumsalz Hydrat

G5422-500G
SIALG5422-500GEA 594 EUR
SIALG5422-500G
di-Natrium-β-glycerophosphat Hydrat, Sigma-Aldrich®
di-Natrium-β-glycerophosphat Hydrat
Formel: C₃H₇Na₂O₆P·H₂O
Molecular Weight: 234,05 g/mol
MDL: MFCD00149082
CAS-Nummer: 154804-51-0
EINECS: 212-464-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR