Girard-Reagenz T (Carbazoylmethyltrimethylammoniumchlorid), Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ200

G900-100G
SIALG900-100GEA 104 EUR
SIALG900-100G
Girard-Reagenz T (Carbazoylmethyltrimethylammoniumchlorid), Sigma-Aldrich®
Girard-Reagenz T (Carbazoylmethyltrimethylammoniumchlorid)
Formel: C₅H₁₄ClN₃O
MDL: MFCD00012009
CAS-Nummer: 123-46-6
EINECS: 204-629-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR