Hydroxylapatit Type I, Suspension, gepuffert, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ200

H0252-25G
SIALH0252-25GEA 182 EUR
SIALH0252-25G
Hydroxylapatit Type I, Suspension, gepuffert, Sigma-Aldrich®
Hydroxylapatit
Formel: Ca₅HO₁₃P₃
MDL: MFCD00010904
CAS-Nummer: 1306-06-5
EINECS: 215-145-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR