Hypoxanthine-Aminopterin-Thymidin (HAT) media supplement, Hybri-Max™
Lieferant: Merck
H0262-10VL
SIALH0262-10VLEA
190
EUR
SIALH0262-10VL
Hypoxanthine-Aminopterin-Thymidin (HAT) media supplement, Hybri-Max™
Medien für die Mikrobiologie
Additive und Supplemente für die Mikrobiologie
Hypoxanthine-Aminopterin-Thymidine (HAT) medium is used for the fusion of myeloma cells with spleen cells and the selection of hybridomas (cell hybrids) in culture.
- Lyophilised powder
- Gamma-irradiated
Supplements are available in vials with 10 ml sterile culture medium, each one sufficient to supplement 500 ml of medium.