Kalium-2,5-dihydroxybenzolsulfonat, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
2,5-Dihydroxybenzenesulphonic acid potassium sal, Hydroquinonesulphonic acid potassium salt, Potassium 2,5-dihydroxybenzenesulphonate
M-Clarity™ Qualitätsstufe = MQ100
H18402-100G
SIALH18402-100GEA
39.3
EUR
SIALH18402-100G
Kalium-2,5-dihydroxybenzolsulfonat, Sigma-Aldrich®
Kalium-2,5-dihydroxybenzolsulfonat
Formel:
C₆H₅KO₅S Molecular Weight: 228,27 g/mol |
MDL:
MFCD00007475 CAS-Nummer: 21799-87-1 EINECS: 244-584-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...