Hydrocortison-21-acetat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Hydrocortisone acetate

M-Clarity™ Qualitätsstufe = MQ200

H3035-5G
SIALH3035-5GEA 158 EUR
SIALH3035-5G
Hydrocortison-21-acetat, Sigma-Aldrich®
Hydrocortison-21-acetat
Formel: C₂₃H₃₂O₆
Molecular Weight: 404,51 g/mol
MDL: MFCD00037714
CAS-Nummer: 50-03-3
EINECS: 200-004-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR