5-Hydroxyindol, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
1H-Indol-5-ol, 5-Indolol, Indol-5-ol, 5-Hydroxy-1H-indol
M-Clarity™ Qualitätsstufe = MQ100
H31859-1G
SIALH31859-1GEA
64.6
EUR
SIALH31859-1G
5-Hydroxyindol, Sigma-Aldrich®
5-Hydroxyindol
Formel:
C₈H₇NO Molecular Weight: 133,15 g/mol Schmelzpunkt: 107…108 °C |
MDL:
MFCD00005677 CAS-Nummer: 1953-54-4 EINECS: 217-782-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...