Heptyl-β-D-thioglucopyranosid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Heptyl thioglucoside, Heptyl-β-D-1-thioglucopyranoside, n-Heptyl β-D-thioglucopyranoside

M-Clarity™ Qualitätsstufe = MQ100

H3264-1G
SIALH3264-1GEA 332 EUR
SIALH3264-1G
Heptyl-β-D-thioglucopyranosid, Sigma-Aldrich®
Heptyl-β-D-thioglucopyranosid
Formel: C₁₃H₂₆O₅S
Molecular Weight: 294,41 g/mol
MDL: MFCD00043236
CAS-Nummer: 85618-20-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR