5-Isochinolinol, Sigma-Aldrich®

Lieferant: Merck
Achtung

Synonyme: 5-Hydroxyisoquinoline

M-Clarity™ Qualitätsstufe = MQ100

H33207-5G
SIALH33207-5GEA 118 EUR
SIALH33207-5G
5-Isochinolinol, Sigma-Aldrich®
5-Isochinolinol
Formel: C₉H₇NO
Molecular Weight: 145.16 g/mol
MDL: MFCD00006906
CAS-Nummer: 2439-04-5
EINECS: 219-456-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR