4'-Hydroxy-3'-methylacetophenon, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

H38004-25G
SIALH38004-25GEA 133 EUR
SIALH38004-25G
4'-Hydroxy-3'-methylacetophenon, Sigma-Aldrich®
4'-Hydroxy-3'-methylacetophenon
Formel: C₉H₁₀O₂
Molecular Weight: 150.18 g/mol
MDL: MFCD00002231
CAS-Nummer: 876-02-8
EINECS: 212-880-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR