N-(2-Hydroxyethyl)iminodiessigsäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2,2'-((2-Hydroxyethyl)azanediyl)diacetic acid

H4271-50G
SIALH4271-50GEA 483 EUR
SIALH4271-50G
N-(2-Hydroxyethyl)iminodiessigsäure, Sigma-Aldrich®
N-(2-Hydroxyethyl)iminodiessigsäure
Formel: C₆H₁₁NO₅
Molecular Weight: 177,155 g/mol
MDL: MFCD00004293
CAS-Nummer: 93-62-9
EINECS: 202-263-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR