3-Hydroxytyrosol ≥98% (durch HPLC), Sigma-Aldrich®

Lieferant: Merck
Achtung

Synonyme: 2-(3,4-Dihydroxyphenyl)ethanol, Hydroxytyrosol, DOPET, Homoprotocatechuyl alcohol, 2-(3,4-Dihydroxyphenyl)ethyl alcohol, 3,4-Dihydroxyphenethyl Alcohol

M-Clarity™ Qualitätsstufe = MQ100

H4291-100MG
SIALH4291-100MGEA 1060 EUR
SIALH4291-100MG
3-Hydroxytyrosol ≥98% (durch HPLC), Sigma-Aldrich®
3-Hydroxytyrosol

Metabolite of oleuropein. Antioxidant. Inhibits the rate of cancer cell proliferation and induces cancer cell apoptosis.

Formel: C₈H₁₀O₃
Molecular Weight: 154,17 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD01320529
CAS-Nummer: 10597-60-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR