5-Hydroxyflavon, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 5-Hydroxy-2-phenyl-4H-benzo[b]pyran-4-on, 5-Hydroxy-2-phenylchromen-4-on, 5-Hydroxy-2-phenyl-4-benzopyron, 5-Hydroxy-2-phenyl-4H-chromen-4-on, 5-Hydroxy-2-phenyl-4H-1-benzopyran-4-on

H4405-250MG
SIALH4405-250MGEA 114 EUR
SIALH4405-250MG
5-Hydroxyflavon, Sigma-Aldrich®
5-Hydroxyflavon
Formel: C₁₅H₁₀O₃
Molecular Weight: 238,24 g/mol
MDL: MFCD00016944
CAS-Nummer: 491-78-1
EINECS: 207-743-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR