7-Hydroxyflavon, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
7-Hydroxy-2-phenyl-4H-1-benzopyran-4-on, 7-Hydroxy-2-phenyl-4-benzopyron, 7-Hydroxy-2-phenyl-4H-benzo[b]pyran-4-on, 7-Hydroxy-2-phenylchromen-4-on, 7-Hydroxy-2-phenyl-4H-chromen-4-on
H4530-1G
SIALH4530-1GEA
54.5
EUR
SIALH4530-1G
7-Hydroxyflavon, Sigma-Aldrich®
7-Hydroxyflavon
Formel:
C₁₅H₁₀O₃ Molecular Weight: 238,24 g/mol Siedepunkt: 395 °C (1013 hPa) Schmelzpunkt: 244…244,5 °C |
MDL:
MFCD00006835 CAS-Nummer: 6665-86-7 EINECS: 229-705-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...