5-Hydroxy-1,4-naphthochinon (Juglon), Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

H47003-1G H47003-5G
SIALH47003-1GEA 66 EUR
SIALH47003-1G SIALH47003-5G
5-Hydroxy-1,4-naphthochinon (Juglon), Sigma-Aldrich®
5-Hydroxy-1,4-naphthochinon (Juglon)
Formel: C₁₀H₆O₃
MDL: MFCD00001684
CAS-Nummer: 481-39-0
EINECS: 207-567-5
UN: 2811
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR