Hydrochlorothiazid, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ200

H4759-5G H4759-100G H4759-25G
SIALH4759-5GEA 72.6 EUR
SIALH4759-5G SIALH4759-100G SIALH4759-25G
Hydrochlorothiazid, Sigma-Aldrich®
Hydrochlorothiazid
Formel: C₇H₈ClN₃O₄S₂
MDL: MFCD00051765
CAS-Nummer: 58-93-5
EINECS: 200-403-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR