Hydrochlorothiazid, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ Qualitätsstufe = MQ200
H4759-5G
H4759-100G
H4759-25G
SIALH4759-5GEA
72.6
EUR
SIALH4759-5G
SIALH4759-100G
SIALH4759-25G
Hydrochlorothiazid, Sigma-Aldrich®
Hydrochlorothiazid
Formel:
C₇H₈ClN₃O₄S₂ |
MDL:
MFCD00051765 CAS-Nummer: 58-93-5 EINECS: 200-403-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...