di-Hydraziniumsulfat, Sigma-Aldrich®

Lieferant: Merck

H4766-100G
SIALH4766-100GEA 44.5 EUR
SIALH4766-100G
di-Hydraziniumsulfat, Sigma-Aldrich®
di-Hydraziniumsulfat
Formel: H₁₀N₄O₄S
Schmelzpunkt: 265 °C
CAS-Nummer: 13464-80-7
EINECS: 236-688-6
UN: 2923
ADR: 8,III
Merck Index: 12,03213

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR