4-Chinazolon, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 4-Hydroxyquinazoline, Quinazolin-4(3H)-one

M-Clarity™ Qualitätsstufe = MQ100

H57807-5G
SIALH57807-5GEA 36 EUR
SIALH57807-5G
4-Chinazolon, Sigma-Aldrich®
4-Chinazolon
Formel: C₈H₆N₂O
Molecular Weight: 146,15 g/mol
MDL: MFCD00511302
CAS-Nummer: 491-36-1
EINECS: 207-735-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR