8-Hydroxy-5-chinolinsulfonsäure Hydrat, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ200

H58757-100G
SIALH58757-100GEA 73.2 EUR
SIALH58757-100G
8-Hydroxy-5-chinolinsulfonsäure Hydrat, Sigma-Aldrich®
8-Hydroxy-5-chinolinsulfonsäure Hydrat
Formel: C₉H₇NO₄S·H₂O
Molecular Weight: 225,22 g/mol (anhy)
MDL: MFCD00149482
CAS-Nummer: 207386-92-3
EINECS: 201-570-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR