cis-4-Hydroxy-D(+)-prolin, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

H5877-10G
SIALH5877-10GEA 816 EUR
SIALH5877-10G
cis-4-Hydroxy-D(+)-prolin, Sigma-Aldrich®
cis-4-Hydroxy-D(+)-prolin
Formel: C₅H₉NO₃
MDL: MFCD00005252
CAS-Nummer: 2584-71-6
EINECS: 219-963-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR