(Hydroxypropyl)methylcellulose (Hypromellose), 4,100 mPa.s (2% in H2O), Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ200

H7509-100G H7509-250G H7509-25G
SIALH7509-100GEA 240 EUR
SIALH7509-100G SIALH7509-250G SIALH7509-25G
(Hydroxypropyl)methylcellulose (Hypromellose), 4,100 mPa.s (2% in H2O), Sigma-Aldrich®
Formel: C₃H₈O₂·xCH₄O
MDL: MFCD00131360
CAS-Nummer: 9004-65-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR