D(-)-Histidin Hydrochlorid Monohydrat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: (R)-(-)-2-Amino-3-(4-imidazoyl)propansäure Hydrochlorid Monohydrat, (R)-(-)-Histidin Hydrochlorid Monohydrat, H-D-His-OH.HCl.H2O, D-3-Imidazol-4-ylalanin Hydrochlorid Monohydrat, (2R)-(-)-Histidin Hydrochlorid Monohydrat

H7625-5G
SIALH7625-5GEA 176 EUR
SIALH7625-5G
D(-)-Histidin Hydrochlorid Monohydrat, Sigma-Aldrich®
D(-)-Histidin Hydrochlorid Monohydrat
Formel: C₆H₉N₃O₂·HCl·1H₂O
Molecular Weight: 209,63 g/mol
Schmelzpunkt: 254 °C
MDL: MFCD00150081
CAS-Nummer: 6341-24-8
EINECS: 228-733-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=848629f46cbda50f344232de988a5ee2dfb98efc&vid=848629f46cbda50f344232de988a5ee2dfb98efc&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR