(Hydroxypropyl)methylcellulose (Hypromellose), M.W. ~26 kDa, 80 - 120 mPa.s (2% in H2O), Millipore®

Lieferant: Merck

H9262-100G H9262-250G H9262-25G
SIALH9262-100GEA 228 EUR
SIALH9262-100G SIALH9262-250G SIALH9262-25G
(Hydroxypropyl)methylcellulose (Hypromellose), M.W. ~26 kDa, 80 - 120 mPa.s (2% in H2O), Millipore®
(Hydroxypropyl)methylcellulose (Hypromellose)
Formel: C₃H₈O₂·xCH₄O
MDL: MFCD00131360
CAS-Nummer: 9004-65-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR