1-(Heptafluorbutyryl)imidazol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 1-(Perfluorobutanoyl)-1H-imidazole, N-Heptafluorobutyrylimidazole

M-Clarity™ Qualitätsstufe = MQ100

H9903-1G
SIALH9903-1GEA 167 EUR
SIALH9903-1G
1-(Heptafluorbutyryl)imidazol, Sigma-Aldrich®
1-(Heptafluorbutyryl)imidazol
Formel: C₇H₃F₇N₂O
Molecular Weight: 264.10 g/mol
MDL: MFCD00014503
CAS-Nummer: 32477-35-3
EINECS: 251-063-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR