D(-)-allo-Isoleucin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: (2R,3S)-(-)-α-Amino-β-methylvaleriansäure, (2R,3S)-(-)-allo-Isoleucin, (2R,3S)-(-)-2-Amino-3-methylpentansäure, H-D-allo-Ile-OH

MQ200

I0380-50MG
SIALI0380-50MGEA 85.4 EUR
SIALI0380-50MG
D(-)-allo-Isoleucin, Sigma-Aldrich®
D(-)-allo-Isoleucin
Formel: C₆H₁₃NO₂
Molecular Weight: 131.17 g/mol
MDL: MFCD00066445
CAS-Nummer: 1509-35-9
EINECS: 216-143-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR