α-Jonon, Sigma-Aldrich®

Lieferant: Merck

Synonyme: (E)-4-(2,6,6-Trimethylcyclohex-2-en-1-yl)but-3-en-2-one

M-Clarity™ Qualitätsstufe = MQ100

I12409-100G
SIALI12409-100GEA 67.1 EUR
SIALI12409-100G
α-Jonon, Sigma-Aldrich®
α-Jonon
Formel: C₁₃H₂₀O
Molecular Weight: 192,3 g/mol
MDL: MFCD00001565
CAS-Nummer: 127-41-3
EINECS: 204-841-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR