β-Jonon, Sigma-Aldrich®

Lieferant: Merck

Synonyme: (E)-4-(2,6,6-Trimethyl-1-cyclohexenyl)-3-buten-2-one, (E)-4-(2,6,6-Trimethylcyclohex-1-en-1-yl)but-3-en-2-one, beta-Ionone

M-Clarity™ Qualitätsstufe = MQ100

I12603-100G
SIALI12603-100GEA 68.3 EUR
SIALI12603-100G
β-Jonon, Sigma-Aldrich®
β-Jonon
Formel: C₁₃H₂₀O
Molecular Weight: 192,30 g/mol
MDL: MFCD00001549
CAS-Nummer: 79-77-6
EINECS: 201-224-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR