Sigma-Aldrich® Intralipid, 20%, emulsion
Lieferant: Merck
Used as a solubilisation agent.
- CAS: 68890-65-3
- MDL: MFCD00287215
Application
Intralipid has been used:
for testing its optical properties to be used as scattering medium for tissue phantom generation
as a component of agarose gel phantom to reduce scatter coefficients and background absorption
in infusion into mice in hyperinsulinemic-euglycemic clamps studies to evaluate its effect on nonesterified free fatty acids (NEFA) circulation
Biochem/physiol Actions:
Intralipid is a lipid emulsion comprising soybean oil. It favors proinflammatory cytokines production and is recommended for the treatment of immune-mediated pregnancy and implantation failure. In parenteral nutrition, intralipid is the source of fats. It elicits protection in heart ischemia-reperfusion injury by inhibiting mitochondrial permeability transition pore opening. Intralipid is an effective alternative for intravenous immunoglobulin and improves bioavailability of platinum based anti-cancer drugs.
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...