N-Isopropylethylendiamin, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
1-Amino-2-((methylethyl)amino)ethan
I22102-1G
SIALI22102-1GEA
25.9
EUR
SIALI22102-1G
N-Isopropylethylendiamin, Sigma-Aldrich®
N-Isopropylethylendiamin
Formel:
C₅H₁₄N₂ Molecular Weight: 102,18 g/mol Siedepunkt: 135 °C (1013 hPa) Dichte: 0,822 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00008164 CAS-Nummer: 19522-67-9 EINECS: 243-125-8 UN: 2733 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...