Indium(III)acetylacetonat, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

I3300-5G
SIALI3300-5GEA 213 EUR
SIALI3300-5G
Indium(III)acetylacetonat, Sigma-Aldrich®
Indium(III)acetylacetonat
Formel: C₁₅H₂₁InO₆
MDL: MFCD00013494
CAS-Nummer: 14405-45-9
EINECS: 238-378-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR