3-Indolylessigsäurehydrazid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Indol-3-ylessigsäurehydrazid, Indol-3-essigsäurehydrazid

M-Clarity™ Qualitätsstufe = MQ100

I3777-10G
SIALI3777-10GEA 319 EUR
SIALI3777-10G
3-Indolylessigsäurehydrazid, Sigma-Aldrich®
3-Indolylessigsäurehydrazid
Formel: C₁₀H₁₁N₃O
Molecular Weight: 189.22 g/mol
Schmelzpunkt: 198
MDL: MFCD00005634
CAS-Nummer: 5448-47-5
EINECS: 226-672-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR