3-Isobutyl-1-methylxanthin, Sigma-Aldrich®
Lieferant: Merck
|
Achtung
|
Synonyme:
3-Isobutyl-1-methyl-2,6(1H,3H)-purindion, 3-Isobutyl-1-methyl-3,7-dihydro-1H-purin-2,6-dion, 3-Isobutyl-1-methyl-3,7-dihydro-2H-purin-2,6-dion, 3,7-Dihydro-3-isobutyl-1-methyl-1H-purin-2,6-dion, 3-Isobutyl-1-methyl-1H,3H-purin-2,6-dion, 2,6-Dihydroxy-1-methyl-3-isobutylpurin
I5879-100MG
I5879-250MG
I5879-1G
I5879-5G
SIALI5879-100MGEA
59.9
EUR
SIALI5879-100MG
SIALI5879-250MG
SIALI5879-1G
SIALI5879-5G
3-Isobutyl-1-methylxanthin, Sigma-Aldrich®
3-Isobutyl-1-methylxanthin
Formel:
C₁₀H₁₄N₄O₂ Schmelzpunkt: 200…201 °C |
MDL:
MFCD00005584 CAS-Nummer: 28822-58-4 EINECS: 249-259-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...