5-Iodcytosin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 4-Amino-5-iod-2-pyrimidinol, 4-Amino-2-hydroxy-5-iodpyrimidin

I6875-5G
SIALI6875-5GEA 1070 EUR
SIALI6875-5G
5-Iodcytosin, Sigma-Aldrich®
5-Iodcytosin
Formel: C₄H₄IN₃O
Molecular Weight: 237 g/mol
MDL: MFCD00023162
CAS-Nummer: 1122-44-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR