5-Iod-2'-deoxyuridin (Idoxuridin), Sigma-Aldrich®

Lieferant: Merck

I7125-25G I7125-5G
SIALI7125-25GEA 1270 EUR
SIALI7125-25G SIALI7125-5G
5-Iod-2'-deoxyuridin (Idoxuridin), Sigma-Aldrich®
5-Iod-2'-deoxyuridin (Idoxuridin)
Formel: C₉H₁₁IN₂O₅
Lagertemperatur: Raumtemperatur
MDL: MFCD00134656
CAS-Nummer: 54-42-2
EINECS: 200-207-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR