5-Iod-2'-deoxyuridin (Idoxuridin), Sigma-Aldrich®
Lieferant: Merck
I7125-25G
I7125-5G
SIALI7125-25GEA
1270
EUR
SIALI7125-25G
SIALI7125-5G
5-Iod-2'-deoxyuridin (Idoxuridin), Sigma-Aldrich®
5-Iod-2'-deoxyuridin (Idoxuridin)
Formel:
C₉H₁₁IN₂O₅ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00134656 CAS-Nummer: 54-42-2 EINECS: 200-207-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...