INT formazan, kristallin, Sigma-Aldrich®

Lieferant: Merck
Gefahr

Synonyme: 1-(4-Iodophenyl)-5-(4-nitrophenyl)-3-phenylformazan, Iodonitrotetrazolium Formazan

M-Clarity™ Qualitätsstufe = MQ100

I7375-1G
SIALI7375-1GEA 206 EUR
SIALI7375-1G
INT formazan, kristallin, Sigma-Aldrich®
INT formazan
Formel: C₁₉H₁₄IN₅O₂
Molecular Weight: 471.26 g/mol
MDL: MFCD00007300
CAS-Nummer: 7781-49-9
EINECS: 231-950-6

Bestellen

Spezifikation Testergebnisse

Infrared spectrum Conforms to structure
Carbon 47.2 - 50.2%
Nitrogen 14.1 - 15.2%
Wavelength 480 - 490 nm
Extinction coefficient (c = 10mg/l; Ethanol) >12000


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR