D(-)-Isoleucin, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
(2R,3R)-(-)-α-Amino-β-methylvaleriansäure, (2R,3R)-(-)-2-Amino-3-methylpentansäure, (2R,3R)-(-)-Isoleucin, H-D-Ile-OH
MQ200
I7634-1G
I7634-250MG
SIALI7634-1GEA
1200
EUR
SIALI7634-1G
SIALI7634-250MG
D(-)-Isoleucin, Sigma-Aldrich®
D(-)-Isoleucin
Formel:
H₃CCH₂CH(CH₃)CH(NH₂)COOH Molecular Weight: 131.17 g/mol Siedepunkt: 235 Schmelzpunkt: 98 Dichte: 1.224 g/cm³ (20 °C) |
MDL:
MFCD00064221 CAS-Nummer: 319-78-8 EINECS: 206-269-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...