4-Iod-L-phenylalanin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: H-Phe(4-I)-OH

M-Clarity™ Qualitätsstufe = MQ200

I8757-1G
SIALI8757-1GEA 274 EUR
SIALI8757-1G
4-Iod-L-phenylalanin, Sigma-Aldrich®
4-Iod-L-phenylalanin
Formel: C₉H₁₀INO₂
Molecular Weight: 291,1 g/mol
MDL: MFCD00002602
CAS-Nummer: 24250-85-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR