(±)-Jasmonsäure, Flüssigkeit, BioReagent pflanzliche Zellkulturen getestet, Sigma-Aldrich®

Lieferant: Merck

Synonyme: (±)-1α,2β-3-Oxo-2-(cis-2-pentenyl)cyclopentaneacetic acid

M-Clarity™ Qualitätsstufe = MQ200

J2500-250MG
SIALJ2500-250MGEA 361 EUR
SIALJ2500-250MG
(±)-Jasmonsäure, Flüssigkeit, BioReagent pflanzliche Zellkulturen getestet, Sigma-Aldrich®
(±)-Jasmonsäure

Jasmonic acid (JA) is a plant hormone derived from linolenic acid. Jasmonic acid is involved in plant cell signaling responses to abiotic and biotic stress.


JA is involved in the regulation of processes that include growth inhibition, senescence, flower development, leaf abscission and tendril coiling. JA is also involved in wound healing of plants and systemic acquired resistance to attack by insects.

Formel: C₁₂H₁₈O₃
Molecular Weight: 210,27 g/mol
MDL: MFCD04221274
CAS-Nummer: 77026-92-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR