Ethyllinoleat, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ200

L1751-5G L1751-1G
SIALL1751-5GEA 197 EUR
SIALL1751-5G SIALL1751-1G
Ethyllinoleat, Sigma-Aldrich®
Ethyllinoleat
Formel: C₂₀H₃₆O₂
Lagertemperatur: Gefrierschrank
MDL: MFCD00009535
CAS-Nummer: 544-35-4
EINECS: 208-868-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR