Bench protectors, Scienceware®, Labmat™
Lieferant: Merck
L2271-1EA
SIALL2271-1EAEA 93.6 EUR
SIALL2271-1EA
Bench protectors, Scienceware®, Labmat™
Oberflächenschutz Oberflächenschutz
Disposable bench and drawer liners protect surfaces from stains and wear.
  • Highly absorbent upper surface traps up to 420 ml/m² of water, dyes, corrosive chemicals or radioactive solutions
  • Flexible polyethylene film backing is non-skid and prevents soak-through of most solutions
  • Rolls are easily cut with a cutter or scissors while roll is retained in dispenser